APOH303C39DF Apollo Scientific Ltd

2-Chloro-5-cyanobenzene-1-sulfonyl chloride

  • MDL number MFCD18392427
  • Cas Number : 942199-56-6
  • Molecular Weight : 236.08

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-chloro-5-cyanobenzenesulfonyl chloride
Inchi Code 1S/C7H3Cl2NO2S/c8-6-2-1-5(4-10)3-7(6)13(9,11)12/h1-3H
Inchi Key IUVMCIXSPYERDI-UHFFFAOYSA-N

Description

Purity 95%
Storage temp. Store at 2-8
Physical Form Solid-Powder
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/2-chloro-5-cyanobenzene-1-sulfonyl-chloride

Safety Information

signalword Danger
pcodes P260;P271;P280
MSDS https://store.apolloscientific.co.uk/product/2-chloro-5-cyanobenzene-1-sulfonyl-chloride
pictograms GHS05;GHS07
hstatements H302+H312+H332;H314
Availability details for
Quantity
  • Availability
  • Add to Cart