APOH303C39F7 Apollo Scientific Ltd

3,6-Difluoro-2-methoxybenzene boronic acid

  • MDL number MFCD11856009
  • Cas Number : 919355-30-9
  • Molecular Weight : 187.94

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3,6-difluoro-2-methoxyphenylboronic acid
Inchi Code 1S/C7H7BF2O3/c1-13-7-5(10)3-2-4(9)6(7)8(11)12/h2-3,11-12H,1H3
Inchi Key AXJIPGDXAIMBDX-UHFFFAOYSA-N

Description

Synonym (3,6-Difluoro-2-methoxyphenyl)boronic acid
Purity 95%
Storage temp. Store at -20
Physical Form Solid
Shipping Temperature Shipped with Ice Packs
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/36-difluoro-2-methoxybenzene-boronic-acid

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/36-difluoro-2-methoxybenzene-boronic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart