APOH303C39FD Apollo Scientific Ltd

7-(Trifluoromethyl)benzo[D]isoxazol-3-amine

  • MDL number MFCD19441875
  • Cas Number : 229623-52-3
  • Molecular Weight : 202.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 7-(trifluoromethyl)-1,2-benzisoxazol-3-amine
Inchi Code 1S/C8H5F3N2O/c9-8(10,11)5-3-1-2-4-6(5)14-13-7(4)12/h1-3H,(H2,12,13)
Inchi Key PIZYYENSQAOYKU-UHFFFAOYSA-N

Description

Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/7-trifluoromethylbenzodisoxazol-3-amine

Safety Information

signalword Danger
pcodes P280;P301+P310;P304+P340
MSDS https://store.apolloscientific.co.uk/product/7-trifluoromethylbenzodisoxazol-3-amine
pictograms GHS06
hstatements H301;H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart