APOH316CDB50 Apollo Scientific Ltd

Ethyl 2-(6-oxo-8-(trifluoromethyl)pyrimido[2,1-b][1,3,5]thiadiazin-3(2H,4H,6H)-yl)acetate

  • MDL number MFCD31812102

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=131314

Safety Information

signalword Warning
pcodes P280;P301+P312;P302+P352
MSDS http://www.apolloscientific.co.uk/display_item.php?id=131314
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart