APOH316CDB51 Apollo Scientific Ltd

2-Chloro-N-((dimethylamino)methylene)-4-fluorobenzamide 250mg pack

  • MDL number MFCD31812107
  • Cas Number : 2379917-15-2
  • Molecular Weight : 228.65

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/2-chloro-n-dimethylaminomethylene-4-fluorobenzamide

Safety Information

signalword Warning
pcodes P280;P301+P312;P304+P340
pictograms GHS07
hstatements H302+H312+H332
Availability details for
Quantity
  • Availability
  • Add to Cart