APOH316CDB56 Apollo Scientific Ltd

1-(8-Chloro-3-(methylsulfonyl)-6-(trifluoromethyl)imidazo[1,5-a]pyridin-1-yl)-2,2,2-trifluoroethanon

  • MDL number MFCD31812112
  • Cas Number : 2379918-52-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/1-8-chloro-3-methylsulfonyl-6-trifluoromethylimidazo15-apyridin-1-yl-222-trifluoroethanone

Safety Information

signalword Warning
pcodes P280;P302+P352;P304+P340
MSDS https://store.apolloscientific.co.uk/product/1-8-chloro-3-methylsulfonyl-6-trifluoromethylimidazo15-apyridin-1-yl-222-trifluoroethanone
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart