APOH316CDB64 Apollo Scientific Ltd

6-Phenyl-1-hexanol

  • MDL number MFCD00014063
  • Cas Number : 2430-16-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Ambient temperature
Physical Form Liquid-Oil
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=131189

Safety Information

signalword Warning
pcodes P280;P302+P352;P304+P340
MSDS http://www.apolloscientific.co.uk/display_item.php?id=131189
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart