APOH316CDB66 Apollo Scientific Ltd

1,4-Dibromo-2-iodobenzene

  • MDL number MFCD07778993
  • Cas Number : 89284-52-6
  • Molecular Weight : 361.8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 38-39
Iupac Name 1,4-dibromo-2-iodobenzene
Inchi Code 1S/C6H3Br2I/c7-4-1-2-5(8)6(9)3-4/h1-3H
Inchi Key VLRYPRKTXDPVNN-UHFFFAOYSA-N

Description

Purity 99%
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 38-39
Product Link https://store.apolloscientific.co.uk/product/14-dibromo-2-iodobenzene

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/14-dibromo-2-iodobenzene
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart