APOH316CDB69 Apollo Scientific Ltd

3'-Iodo-2'-methylacetophenone

  • MDL number MFCD22573017
  • Cas Number : 52164-39-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 1-(3-Iodo-2-methylphenyl)ethan-1-one)
Purity 95
Storage temp. Ambient temperature
Physical Form Liquid-Oil
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=125977

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=125977
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart