APOH316CDB6B Apollo Scientific Ltd

2,6-Diethylphenyl isocyanate

  • MDL number MFCD00019912
  • Cas Number : 20458-99-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Physical Form Liquid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=129725

Safety Information

signalword Danger
pcodes P260;P301+P310;P284;P362+P364
MSDS http://www.apolloscientific.co.uk/display_item.php?id=129725
pictograms GHS06;GHS08
hstatements H301;H315;H319;H330;H335
Availability details for
Quantity
  • Availability
  • Add to Cart