APOH316CDB71 Apollo Scientific Ltd

2-Bromo-N,N-dimethylacetamide

  • MDL number MFCD12026683
  • Cas Number : 5468-77-9
  • Molecular Weight : 166.02

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-bromo-N,N-dimethylacetamide
density 1.5412
Inchi Code 1S/C4H8BrNO/c1-6(2)4(7)3-5/h3H2,1-2H3
Inchi Key QPIOVNJLOVNTMW-UHFFFAOYSA-N

Description

Purity 98%
Density 1.5412
Flash Point 63-65/1mm
Storage temp. Store at 2-8
Physical Form Liquid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/2-bromo-nn-dimethylacetamide

Safety Information

signalword Danger
pcodes P260;P271;P280
MSDS https://store.apolloscientific.co.uk/product/2-bromo-nn-dimethylacetamide
pictograms GHS05
hstatements H314
Availability details for
Quantity
  • Availability
  • Add to Cart