APOH316CDB72 Apollo Scientific Ltd

N-Phenylpyridine-3-carboxamide

  • MDL number MFCD00023572
  • Cas Number : 1752-96-1
  • Molecular Weight : 198.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 119-120
Iupac Name N-phenylnicotinamide
Inchi Code 1S/C12H10N2O/c15-12(10-5-4-8-13-9-10)14-11-6-2-1-3-7-11/h1-9H,(H,14,15)
Inchi Key NYQXIOZBHWFCBU-UHFFFAOYSA-N

Description

Purity 95%
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 119-120
Product Link https://store.apolloscientific.co.uk/product/n-phenylpyridine-3-carboxamide

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/n-phenylpyridine-3-carboxamide
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart