APOH316CDB75 Apollo Scientific Ltd

3-Iodo-2-methylbenzyl bromide

  • MDL number MFCD18397827
  • Cas Number : 950577-16-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 3-(Bromomethyl)-2-methyliodobenzene
Purity 95
Storage temp. Ambient temperature
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=125401

Safety Information

signalword Danger
pcodes P271;P260;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=125401
pictograms GHS05
hstatements H314
Availability details for
Quantity
  • Availability
  • Add to Cart