APOH316CDB76 Apollo Scientific Ltd

3-Iodo-2-methylbenzaldehyde

  • MDL number MFCD18397774
  • Cas Number : 848444-83-7
  • Molecular Weight : 246.05

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Physical Form Solid-Powder
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/3-iodo-2-methylbenzaldehyde

Safety Information

signalword Warning
pcodes P280;P301+P312;P304+P340
MSDS https://store.apolloscientific.co.uk/product/3-iodo-2-methylbenzaldehyde
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart