APOH316CDB78 Apollo Scientific Ltd

N-3-Thienylacetamide

  • MDL number MFCD00159570
  • Cas Number : 42602-67-5
  • Molecular Weight : 141.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name N-(3-thienyl)acetamide
Inchi Code 1S/C6H7NOS/c1-5(8)7-6-2-3-9-4-6/h2-4H,1H3,(H,7,8)
Inchi Key QHXQVXGAEYDJCR-UHFFFAOYSA-N

Description

Purity 99%
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/n-3-thienylacetamide

Safety Information

signalword Warning
pcodes P280;P301+P312;P304+P340
MSDS https://store.apolloscientific.co.uk/product/n-3-thienylacetamide
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart