APOH316CDB7B Apollo Scientific Ltd

2-Chloro-4-nitrophenyl isocyanate

  • MDL number MFCD00037063
  • Cas Number : 40397-95-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 98
Storage temp. Ambient temperature
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=129726

Safety Information

signalword Danger
pcodes P271;P261;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=129726
pictograms GHS07;GHS08
hstatements H315;H317;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart