APOH316CDB7C Apollo Scientific Ltd

Methyl 5-iodo-2-methylbenzoate

  • MDL number MFCD11112847
  • Cas Number : 103440-54-6
  • Molecular Weight : 276.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

bp 60
Iupac Name methyl 5-iodo-2-methylbenzoate
Inchi Code 1S/C9H9IO2/c1-6-3-4-7(10)5-8(6)9(11)12-2/h3-5H,1-2H3
Inchi Key BXVIKPGEGLOGLU-UHFFFAOYSA-N

Description

Purity 95%
Storage temp. Store at 2-8
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Boiling point 60
Product Link https://store.apolloscientific.co.uk/product/methyl-5-iodo-2-methylbenzoate

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/methyl-5-iodo-2-methylbenzoate
pictograms GHS07
hstatements H302;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart