APOH316CDB81 Apollo Scientific Ltd

2-(Cyclopropyl)benzyl alcohol

  • MDL number MFCD20921818
  • Cas Number : 118184-68-2
  • Molecular Weight : 148.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/2-cyclopropylbenzyl-alcohol

Safety Information

signalword Warning
pcodes P271;P261;P280
MSDS https://store.apolloscientific.co.uk/product/2-cyclopropylbenzyl-alcohol
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart