APOH316CDB84 Apollo Scientific Ltd

3-(Cyclopropyl)benzyl alcohol

  • MDL number MFCD06802678
  • Cas Number : 893738-74-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Physical Form Liquid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link https://store.apolloscientific.co.uk/product/3-cyclopropylbenzyl-alcohol

Safety Information

signalword Warning
pcodes P280
MSDS https://store.apolloscientific.co.uk/product/3-cyclopropylbenzyl-alcohol
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart