APOH316CDB86 Apollo Scientific Ltd

Diethyl 2-oxobutanedioate

  • MDL number MFCD00511083
  • Cas Number : 108-56-5
  • Molecular Weight : 188.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name diethyl 2-oxosuccinate
density 1.14
Inchi Code 1S/C8H12O5/c1-3-12-7(10)5-6(9)8(11)13-4-2/h3-5H2,1-2H3
Inchi Key JDXYSCUOABNLIR-UHFFFAOYSA-N

Description

Synonym Diethyl oxalacetate
Purity 95%
Density 1.14
Flash Point 87
Storage temp. Store at -20
Physical Form Liquid
Shipping Temperature Shipped with Ice Packs
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/diethyl-2-oxobutanedioate

Safety Information

signalword Warning
pcodes P280
MSDS https://store.apolloscientific.co.uk/product/diethyl-2-oxobutanedioate
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart