APOH316CDB88 Apollo Scientific Ltd

3-(Cyclopropyl)benzyl bromide

  • MDL number MFCD15527185
  • Cas Number : 1260850-05-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Storage temp. Ambient temperature
Physical Form Liquid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=125825

Safety Information

signalword Danger
pcodes P260;P280;P310
MSDS http://www.apolloscientific.co.uk/display_item.php?id=125825
pictograms GHS05;GHS07
hstatements H302+H312+H332;H314
Availability details for
Quantity
  • Availability
  • Add to Cart