APOH316CDB89 Apollo Scientific Ltd

Methyl 5-(bromomethyl)-2-methylbenzoate

  • MDL number MFCD28148164
  • Cas Number : 501362-17-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Ambient temperature
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=125603

Safety Information

signalword Danger
pcodes P271;P260;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=125603
pictograms GHS05
hstatements H314
Availability details for
Quantity
  • Availability
  • Add to Cart