APOH316CDB8B Apollo Scientific Ltd

4-Bromo-1-benzofuran-2-carboxylic acid

  • MDL number MFCD22034498
  • Cas Number : 1369143-28-1
  • Molecular Weight : 241.04

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 4-bromo-1-benzofuran-2-carboxylic acid
Inchi Code 1S/C9H5BrO3/c10-6-2-1-3-7-5(6)4-8(13-7)9(11)12/h1-4H,(H,11,12)
Inchi Key AWHJIEGQJDBVCF-UHFFFAOYSA-N

Description

Storage temp. Store at 2-8
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/4-bromo-1-benzofuran-2-carboxylic-acid

Safety Information

signalword Warning
pcodes P280;P304+P340
MSDS https://store.apolloscientific.co.uk/product/4-bromo-1-benzofuran-2-carboxylic-acid
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart