APOH316CDB8E Apollo Scientific Ltd

4-(Cyclopropyl)benzyl bromide

  • MDL number MFCD12024391
  • Cas Number : 1150617-57-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Storage temp. Ambient temperature
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=125824

Safety Information

signalword Danger
pcodes P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=125824
pictograms GHS05;GHS07
hstatements H314
Availability details for
Quantity
  • Availability
  • Add to Cart