APOH316CDB8F Apollo Scientific Ltd

2-(Cyclopropyl)benzyl bromide

  • MDL number MFCD15527184
  • Cas Number : 1260801-90-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Storage temp. Ambient temperature
Physical Form Liquid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=125826

Safety Information

signalword Danger
pcodes P280;P301+P312;P301+P330+P331
MSDS http://www.apolloscientific.co.uk/display_item.php?id=125826
pictograms GHS05;GHS07
hstatements H302+H312+H332;H314
Availability details for
Quantity
  • Availability
  • Add to Cart