APOH316CDB92 Apollo Scientific Ltd

3-(Bromomethyl)aniline, N-BOC protected

  • MDL number MFCD11974927
  • Cas Number : 118684-32-5
  • Molecular Weight : 286.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/3-bromomethylaniline-n-boc-protected

Safety Information

signalword Danger
pcodes P271;P260;P280
MSDS https://store.apolloscientific.co.uk/product/3-bromomethylaniline-n-boc-protected
pictograms GHS05;GHS07
hstatements H302+H312+H332;H314
Availability details for
Quantity
  • Availability
  • Add to Cart