APOH316CDB9C Apollo Scientific Ltd

Ethyl 5-bromo-7-methoxy-1-benzofuran-2-carboxylate

  • MDL number MFCD16089983
  • Cas Number : 150612-67-2
  • Molecular Weight : 299.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name ethyl 5-bromo-7-methoxy-1-benzofuran-2-carboxylate
Inchi Code 1S/C12H11BrO4/c1-3-16-12(14)10-5-7-4-8(13)6-9(15-2)11(7)17-10/h4-6H,3H2,1-2H3
Inchi Key GDKMBFBGOSPMPD-UHFFFAOYSA-N

Description

Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/ethyl-5-bromo-7-methoxy-1-benzofuran-2-carboxylate

Safety Information

signalword Warning
pcodes P301+P312;P305+P351+P338
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart