APOH316CDBA0 Apollo Scientific Ltd

5-Iodo-3,4-dihydro-2H-1,8-naphthyridine, N1-BOC protected

  • MDL number MFCD31745691

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym tert-Butyl 5-iodo-3,4-dihydro-2H-1,8-naphthyridine-1-carboxylate
Purity 95
Storage temp. Ambient temperature
Physical Form Solid-Powder
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=128633

Safety Information

signalword Warning
pcodes P280;P301+P312
MSDS http://www.apolloscientific.co.uk/display_item.php?id=128633
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart