APOH316CDBA3 Apollo Scientific Ltd

(2S)-4-Iodo-2-isopropyl-2,3-dihydro-1H-pyrrolo[2,3-b]pyridine

  • MDL number MFCD31793561
  • Cas Number : 2366997-22-8
  • Molecular Weight : 288.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (S)-4-iodo-2-isopropyl-2,3-dihydro-1H-pyrrolo[2,3-b]pyridine
Inchi Code 1S/C10H13IN2/c1-6(2)9-5-7-8(11)3-4-12-10(7)13-9/h3-4,6,9H,5H2,1-2H3,(H,12,13)/t9-/m0/s1
Inchi Key XBPOEVXPAJJNMT-VIFPVBQESA-N

Description

Purity 95%
Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/2s-4-iodo-2-isopropyl-23-dihydro-1h-pyrrolo23-bpyridine

Safety Information

signalword Warning
pcodes P280;P301+P312
MSDS https://store.apolloscientific.co.uk/product/2s-4-iodo-2-isopropyl-23-dihydro-1h-pyrrolo23-bpyridine
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart