APOH316CDBA4 Apollo Scientific Ltd

(2S)-4-Iodo-2-methyl-2,3-dihydropyrrolo[2,3-b]pyridine, N-BOC protected

  • Cas Number : 2366997-11-5
  • Molecular Weight : 360.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name tert-butyl (S)-4-iodo-2-methyl-2,3-dihydro-1H-pyrrolo[2,3-b]pyridine-1-carboxylate
Inchi Code 1S/C13H17IN2O2/c1-8-7-9-10(14)5-6-15-11(9)16(8)12(17)18-13(2,3)4/h5-6,8H,7H2,1-4H3/t8-/m0/s1
Inchi Key OGTRGABAZMACSR-QMMMGPOBSA-N

Description

Purity 95%
Storage temp. Ambient Temperature
Physical Form Liquid-Oil
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/2s-4-iodo-2-methyl-23-dihydropyrrolo23-bpyridine-n-boc-protected

Safety Information

signalword Warning
pcodes P280;P301+P312;P362+P364
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart