APOH316CDBAB Apollo Scientific Ltd

3-Iodo-4-methylbenzyl bromide

  • MDL number MFCD09745120
  • Cas Number : 626250-55-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=128486

Safety Information

MSDS http://www.apolloscientific.co.uk/display_item.php?id=128486
Availability details for
Quantity
  • Availability
  • Add to Cart