APOH316CDBAE Apollo Scientific Ltd

5-Chlorothieno[3,2-b]pyridine-6-carbaldehyde

  • MDL number MFCD18838815
  • Cas Number : 65977-56-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 99
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=125375

Safety Information

MSDS http://www.apolloscientific.co.uk/display_item.php?id=125375
Availability details for
Quantity
  • Availability
  • Add to Cart