APOH316CDBAF Apollo Scientific Ltd

(5-Methyl-1,3-oxazol-4-yl)methanol

  • MDL number MFCD16556111
  • Cas Number : 874821-67-7
  • Molecular Weight : 113.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (5-methyl-1,3-oxazol-4-yl)methanol
Inchi Code 1S/C5H7NO2/c1-4-5(2-7)6-3-8-4/h3,7H,2H2,1H3
Inchi Key FKQJNZYJRXRLNZ-UHFFFAOYSA-N

Description

Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/5-methyl-13-oxazol-4-ylmethanol

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/5-methyl-13-oxazol-4-ylmethanol
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart