APOH316CDBB3 Apollo Scientific Ltd

Ethyl 5-(4-methoxyphenyl)isoxazole-3-carboxylate

  • MDL number MFCD08446406
  • Cas Number : 925006-96-8
  • Molecular Weight : 247.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/ethyl-5-4-methoxyphenylisoxazole-3-carboxylate

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/ethyl-5-4-methoxyphenylisoxazole-3-carboxylate
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart