APOH316CDBB4 Apollo Scientific Ltd

Ethyl 6-chlorobenzofuran-2-carboxylate

  • MDL number MFCD28987315
  • Cas Number : 74222-19-8
  • Molecular Weight : 224.64

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/ethyl-6-chlorobenzofuran-2-carboxylate

Safety Information

signalword Warning
pcodes P280;P304+P340
MSDS https://store.apolloscientific.co.uk/product/ethyl-6-chlorobenzofuran-2-carboxylate
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart