APOH316CDBB6 Apollo Scientific Ltd

7-Bromobenzofuran-2-carboxylic acid

  • MDL number MFCD10699647
  • Cas Number : 550998-59-9
  • Molecular Weight : 241.04

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 7-bromo-1-benzofuran-2-carboxylic acid
Inchi Code 1S/C9H5BrO3/c10-6-3-1-2-5-4-7(9(11)12)13-8(5)6/h1-4H,(H,11,12)
Inchi Key OPYANDYWSZSRSD-UHFFFAOYSA-N

Description

Storage temp. Ambient Temperature
Physical Form Solid
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/7-bromobenzofuran-2-carboxylic-acid

Safety Information

signalword Warning
pcodes P280;P301+P312
MSDS https://store.apolloscientific.co.uk/product/7-bromobenzofuran-2-carboxylic-acid
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart