APOH316CDBBA Apollo Scientific Ltd

tert-Butyl (2R)-4-iodo-2-isopropyl-2,3-dihydropyrrolo[2,3-b]pyridine-1-carboxylate

  • Cas Number : 2366996-94-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Ambient temperature
Physical Form Solid-Powder
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=130342

Safety Information

signalword Warning
pcodes P280;P301+P312
MSDS http://www.apolloscientific.co.uk/display_item.php?id=130342
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart