APOH316CDBBD Apollo Scientific Ltd

(2R)-4-Iodo-2-methyl-2,3-dihydro-1H-pyrrolo[2,3-b]pyridine, N-BOC protected

  • Cas Number : 2366997-18-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Ambient temperature
Physical Form Liquid-Oil
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=129395

Safety Information

signalword Warning
pcodes P280;P301+P312;P302+P352
MSDS http://www.apolloscientific.co.uk/display_item.php?id=129395
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart