APOH316CDBC1 Apollo Scientific Ltd

(2S)-4-Iodo-2-methyl-2,3-dihydro-1H-pyrrolo[2,3-b]pyridine

  • Cas Number : 2366997-19-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Ambient temperature
Physical Form Solid-Powder
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=129546

Safety Information

signalword Warning
pcodes P280;P301+P312
MSDS http://www.apolloscientific.co.uk/display_item.php?id=129546
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart