APOH316CDBC5 Apollo Scientific Ltd

6-Chlorobenzofuran

  • MDL number MFCD18825837
  • Cas Number : 151619-12-4
  • Molecular Weight : 152.58

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 6-chloro-1-benzofuran
Inchi Code 1S/C8H5ClO/c9-7-2-1-6-3-4-10-8(6)5-7/h1-5H
Inchi Key BKBDOWJPNZYBSO-UHFFFAOYSA-N

Description

Storage temp. Ambient Temperature
Shipping Temperature Ambient Temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/6-chlorobenzofuran

Safety Information

signalword Warning
pcodes P261;P271;P280
MSDS https://store.apolloscientific.co.uk/product/6-chlorobenzofuran
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart