APOH316CDBD1 Apollo Scientific Ltd

3-Ethoxyacrylic acid

  • MDL number MFCD01566841
  • Cas Number : 6192-01-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 109-111
Synonym 3-Ethoxyprop-2-enoic acid
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Boiling point 87/5mm
Product Link https://store.apolloscientific.co.uk/product/3-ethoxyacrylic-acid

Safety Information

signalword Danger
pcodes P271;P260;P280
MSDS https://store.apolloscientific.co.uk/product/3-ethoxyacrylic-acid
pictograms GHS05
hstatements H314
Availability details for
Quantity
  • Availability
  • Add to Cart