APOH316CDBD5 Apollo Scientific Ltd

3-Aminomethylisoxazole

  • MDL number MFCD06738791
  • Cas Number : 131052-58-9
  • Molecular Weight : 98.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym 1,2-Oxazol-3-ylmethanamine
Purity 95
Physical Form Liquid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/3-aminomethylisoxazole

Safety Information

signalword Danger
pcodes P260;P271;P280
MSDS https://store.apolloscientific.co.uk/product/3-aminomethylisoxazole
pictograms GHS05
hstatements H314
Availability details for
Quantity
  • Availability
  • Add to Cart