APOH316CDBD8 Apollo Scientific Ltd

Bis(isopropyl) azodicarboxylate

  • MDL number MFCD00008875
  • Cas Number : 2446-83-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 43223
Synonym DIAD, Diprop-2-yl diazene-1,2-dicarboxylate
Purity 98
Density 1.027
Flash Point 106
Storage temp. Store at 2-8
Physical Form Liquid
Shipping Temperature Ship at ambient temperature
Country of Origin United Kingdom
Boiling point 74-75/0.3mmHg
Product Link http://www.apolloscientific.co.uk/display_item.php?id=28607

Safety Information

signalword Warning
pcodes P260;P271;P280
MSDS http://www.apolloscientific.co.uk/display_item.php?id=28607
pictograms GHS07;GHS08;GHS09
hstatements H319;H335;H411
Availability details for
Quantity
  • Availability
  • Add to Cart