APOH316CDBE4 Apollo Scientific Ltd

2,4-Dimethylbenzyl alcohol

  • MDL number MFCD00004624
  • Cas Number : 16308-92-2
  • Molecular Weight : 136.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 25
Purity 97
Physical Form Solid-low melt
Shipping Temperature Ambient temperature
Country of Origin GB
Boiling point 120
Product Link https://store.apolloscientific.co.uk/product/24-dimethylbenzyl-alcohol

Safety Information

signalword Warning
pcodes P280;P302+P352
MSDS https://store.apolloscientific.co.uk/product/24-dimethylbenzyl-alcohol
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart