APOH316CDC39 Apollo Scientific Ltd

Sodium 4-chlorobenzenesulfonate

  • MDL number MFCD00014731
  • Cas Number : 5138-90-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Synonym Sodium 4-chlorobenzenesulphonate
Purity 98
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=122484

Safety Information

MSDS http://www.apolloscientific.co.uk/display_item.php?id=122484
Availability details for
Quantity
  • Availability
  • Add to Cart