APOH316CDD0C Apollo Scientific Ltd

2-(4-Chlorophenyl)-2-oxoacetaldehyde

  • MDL number MFCD03411516
  • Cas Number : 4996-21-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Storage temp. Ambient temperature
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=131328

Safety Information

signalword Warning
pcodes P280;P304+P340
MSDS http://www.apolloscientific.co.uk/display_item.php?id=131328
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart