APOH316CDD0D Apollo Scientific Ltd

Methyl hydrazinecarbodithioate

  • MDL number MFCD00463356
  • Cas Number : 5397-03-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=131310

Safety Information

MSDS http://www.apolloscientific.co.uk/display_item.php?id=131310
Availability details for
Quantity
  • Availability
  • Add to Cart