APOH316CDD0F Apollo Scientific Ltd

1-(4-Bromophenyl)pyrrolidine

  • MDL number MFCD04112480
  • Cas Number : 22090-26-2
  • Molecular Weight : 226.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 107
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin GB
Product Link https://store.apolloscientific.co.uk/product/1-4-bromophenylpyrrolidine

Safety Information

signalword Warning
pcodes P280;P305+P351+P338
MSDS https://store.apolloscientific.co.uk/product/1-4-bromophenylpyrrolidine
pictograms GHS07
hstatements H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart