APOH316CDD10 Apollo Scientific Ltd

6-Methoxyspiro[benzo[e][1,3]oxazine-2,4'-piperidin]-4(3H)-one hydrochloride

  • MDL number MFCD22372630
  • Cas Number : 1204346-31-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=131308

Safety Information

MSDS http://www.apolloscientific.co.uk/display_item.php?id=131308
Availability details for
Quantity
  • Availability
  • Add to Cart