APOH316CDD1C Apollo Scientific Ltd

4-(Benzylamino)furan-2(5H)-one

  • MDL number MFCD03005353
  • Cas Number : 38470-54-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Storage temp. Ambient temperature
Physical Form Solid
Shipping Temperature Ambient temperature
Country of Origin United Kingdom
Product Link http://www.apolloscientific.co.uk/display_item.php?id=131290

Safety Information

signalword Warning
pcodes P280;P301+P312;P304+P340
MSDS http://www.apolloscientific.co.uk/display_item.php?id=131290
pictograms GHS07
hstatements H302+H312+H332;H315;H319;H335
Availability details for
Quantity
  • Availability
  • Add to Cart